6-(4-Chlorophenoxy)pyridin-3-amine
Supplier: Apollo Scientific
OR111073-10G
OR111073-1G
OR111073-5G
APOSOR111073-10GEA
1360
EUR
APOSOR111073-10G
APOSOR111073-1G
APOSOR111073-5G
6-(4-Chlorophenoxy)pyridin-3-amine
6-(4-Chlorophenoxy)pyridin-3-amine
Formula:
C₁₁H₉ClN₂O MW: 220,655 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00052646 CAS Number: 75926-64-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...