2-(2-Ethylphenyl)hydrazinecarbothioamide

Supplier: Apollo Scientific

OR111085-5G OR111085-10G OR111085-1G
APOSOR111085-5GEA 0 EUR
APOSOR111085-5G APOSOR111085-10G APOSOR111085-1G
2-(2-Ethylphenyl)hydrazinecarbothioamide
2-(2-Ethylphenyl)hydrazinecarbothioamide
Formula: C₉H₁₃N₃S
MW: 195,285 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01566654
CAS Number: 65078-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR