2,4-Dichloro-5-chlorosulphonyl-benzoic acid
Supplier: Apollo Scientific
|
Danger
|
OR111098-1G
OR111098-5G
APOSOR111098-1GEA
94.5
EUR
APOSOR111098-1G
APOSOR111098-5G
2,4-Dichloro-5-chlorosulphonyl-benzoic acid
2,4-Dichloro-5-chlorosulphonyl-benzoic acid
Formula:
C₇H₃Cl₃O₄S MW: 289,52 g/mol Melting Pt: 178…180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00486543 CAS Number: 3740-18-9 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...