N'-(4-Chloro-3-nitrophenyl)-N,N-diisopropylurea

Supplier: Apollo Scientific

OR111134-1G OR111134-5G
APOSOR111134-1GEA 0 EUR
APOSOR111134-1G APOSOR111134-5G
N'-(4-Chloro-3-nitrophenyl)-N,N-diisopropylurea
N'-(4-Chloro-3-nitrophenyl)-N,N-diisopropylurea
Formula: C₁₃H₁₈ClN₃O₃
Storage Temperature: Ambient
MDL Number: MFCD28987631
CAS Number: 1980053-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR