1-(1-Adamantyl)ethanol

Supplier: Apollo Scientific
Warning

OR111208-10G OR111208-1G OR111208-5G
APOSOR111208-10GEA 0 EUR
APOSOR111208-10G APOSOR111208-1G APOSOR111208-5G
1-(1-Adamantyl)ethanol
1-(1-Adamantyl)ethanol
Formula: C₁₂H₂₀O
MW: 180,287 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01321095
CAS Number: 26750-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR