4-Methoxy-N,2,3,6-tetramethylbenzenesulphonamide
Supplier: Apollo Scientific
OR111379-1G
OR111379-5G
APOSOR111379-1GEA
0
EUR
APOSOR111379-1G
APOSOR111379-5G
4-Methoxy-N,2,3,6-tetramethylbenzenesulphonamide
4-Methoxy-N,2,3,6-tetramethylbenzenesulphonamide
Formula:
C₁₁H₁₇NO₃S Storage Temperature: Ambient |
MDL Number:
MFCD30481155 CAS Number: 2197052-81-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...