2-(2-Bromo-4-methylphenyl)acetonitrile

Supplier: Apollo Scientific

OR111407-1G OR111407-250MG
APOSOR111407-1GEA 510 EUR
APOSOR111407-1G APOSOR111407-250MG
2-(2-Bromo-4-methylphenyl)acetonitrile
2-(2-Bromo-4-methylphenyl)acetonitrile
Formula: C₉H₈BrN
MW: 210,071 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18392226
CAS Number: 1069114-80-2
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR