2-(2-Chloro-3,4-dimethoxyphenyl)acetic acid

Supplier: Apollo Scientific

OR111438-1G OR111438-250MG
APOSOR111438-1GEA 284 EUR
APOSOR111438-1G APOSOR111438-250MG
2-(2-Chloro-3,4-dimethoxyphenyl)acetic acid
2-(2-Chloro-3,4-dimethoxyphenyl)acetic acid
Formula: C₁₀H₁₁ClO₄
MW: 230,645 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21606658
CAS Number: 6834-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR