8-Chloro-4H-thieno[3,2-c]thiochromene-2-carboxylic acid
Supplier: Apollo Scientific
|
Warning
|
OR111468-1G
OR111468-250MG
APOSOR111468-1GEA
16.1
EUR
APOSOR111468-1G
APOSOR111468-250MG
8-Chloro-4H-thieno[3,2-c]thiochromene-2-carboxylic acid
8-Chloro-4H-thieno[3,2-c]thiochromene-2-carboxylic acid
Formula:
C₁₂H₇ClO₂S₂ MW: 282,766 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00107150 CAS Number: 255395-56-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...