6-Cyclopropyl-2-hydroxynicotinamide

Supplier: Apollo Scientific
Warning

OR111581-1G OR111581-250MG
APOSOR111581-1GEA 78.3 EUR
APOSOR111581-1G APOSOR111581-250MG
6-Cyclopropyl-2-hydroxynicotinamide
6-Cyclopropyl-2-hydroxynicotinamide
Formula: C₉H₁₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD32066837
CAS Number: 1950617-91-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR