4-Iodoquinoline 97%
Supplier: Apollo Scientific
|
Warning
|
OR11294-1G
OR11294-250MG
APOSOR11294-1GEA
98.6
EUR
APOSOR11294-1G
APOSOR11294-250MG
4-Iodoquinoline 97%
4-Iodoquinoline
Formula:
C₉H₆IN Melting Pt: 94…96 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07700252 CAS Number: 16560-43-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...