(tert-Butyl) 6-bromonicotinate 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(tert-Butyl) 6-bromopyridine-3-carboxylate, tert-Butyl 6-bromonicotinate
OR11518-1G
OR11518-500MG
APOSOR11518-1GEA
176
EUR
APOSOR11518-1G
APOSOR11518-500MG
(tert-Butyl) 6-bromonicotinate 98%
(tert-Butyl) 6-bromonicotinate
Formula:
C₁₀H₁₂BrNO₂ MW: 258,112 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09264580 CAS Number: 941294-58-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...