5-Chloro-2-methoxy-3-nitropyridine
Supplier: Apollo Scientific
|
Warning
|
OR11978-1G
OR11978-5G
APOSOR11978-1GEA
75.7
EUR
APOSOR11978-1G
APOSOR11978-5G
5-Chloro-2-methoxy-3-nitropyridine
5-Chloro-2-methoxy-3-nitropyridine
Formula:
C₆H₅ClN₂O₃ MW: 188,569 g/mol Melting Pt: 90 °C Storage Temperature: Ambient |
MDL Number:
MFCD07375032 CAS Number: 22353-52-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...