4-Bromo-3-(hydroxymethyl)-1-methyl-1H-pyrazole 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(4-Bromo-1-methyl-1H-pyrazol-3-yl)methanol
OR12252-1G
OR12252-5G
OR12252-10G
APOSOR12252-1GEA
34.8
EUR
APOSOR12252-1G
APOSOR12252-5G
APOSOR12252-10G
4-Bromo-3-(hydroxymethyl)-1-methyl-1H-pyrazole 97%
4-Bromo-3-(hydroxymethyl)-1-methyl-1H-pyrazole
Formula:
C₅H₇BrN₂O MW: 191,026 g/mol Melting Pt: 94…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD06202888 CAS Number: 915707-65-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...