2-Cyclohexylpyrrolidine

Supplier: Apollo Scientific
Warning

OR12518-100MG OR12518-500MG OR12518-1G
APOSOR12518-100MGEA 261 EUR
APOSOR12518-100MG APOSOR12518-500MG APOSOR12518-1G
2-Cyclohexylpyrrolidine
2-Cyclohexylpyrrolidine
Formula: C₁₀H₁₉N
MW: 153.265 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631841
CAS Number: 367281-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR