Biphenyl-2-sulphonyl chloride 95%

Supplier: Apollo Scientific
Danger

Synonyms: 2-Phenylbenzenesulphonyl chloride, Biphenyl-2-sulphonyl chloride

OR12793-100MG OR12793-1G OR12793-250MG
APOSOR12793-100MGEA 58.2 EUR
APOSOR12793-100MG APOSOR12793-1G APOSOR12793-250MG
Biphenyl-2-sulphonyl chloride 95%
Biphenyl-2-sulphonyl chloride
Formula: C₁₂H₉ClO₂S
MW: 252,717 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02246591
CAS Number: 2688-90-6
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR