2-(2-Methylphenoxy)benzoic acid 95%

Supplier: Apollo Scientific
Warning

Synonyms: 2-(o-Tolyloxy)benzoic acid

OR12944-1G OR12944-5G
APOSOR12944-1GEA 506 EUR
APOSOR12944-1G APOSOR12944-5G
2-(2-Methylphenoxy)benzoic acid 95%
2-(2-Methylphenoxy)benzoic acid
Formula: C₁₄H₁₂O₃
MW: 228,243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00439674
CAS Number: 6325-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR