2-Methyl-1,3-benzoxazole-7-carboxylic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
7-Carboxy-2-methylbenzo[d]oxazole, 7-Carboxy-2-methyl-1,3-benzoxazole
OR13439-100MG
OR13439-1G
OR13439-250MG
APOSOR13439-100MGEA
109
EUR
APOSOR13439-100MG
APOSOR13439-1G
APOSOR13439-250MG
2-Methyl-1,3-benzoxazole-7-carboxylic acid
2-Methyl-1,3-benzoxazole-7-carboxylic acid
Formula:
C₉H₇NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD03308601 CAS Number: 52395-92-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...