Ethyl 6-amino-5-bromonicotinate 95%

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 2-amino-3-bromo-5-pyridinecarboxylate

OR1349-1G OR1349-250MG
APOSOR1349-1GEA 23.4 EUR
APOSOR1349-1G APOSOR1349-250MG
Ethyl 6-amino-5-bromonicotinate 95%
Ethyl 6-amino-5-bromonicotinate
Formula: C₈H₉BrN₂O₂
MW: 245,073 g/mol
Melting Pt: 125…127 °C
Storage Temperature: Ambient
MDL Number: MFCD06659894
CAS Number: 850429-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR