4-[4-(Hydroxymethyl)phenyl]pyridine 95+%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
(4-Pyridin-4-ylphenyl)methanol, (4-(Pyridin-4-yl)phenyl)methanol
OR1432-1G
OR1432-250MG
APOSOR1432-1GEA
203
EUR
APOSOR1432-1G
APOSOR1432-250MG
4-[4-(Hydroxymethyl)phenyl]pyridine 95+%
4-[4-(Hydroxymethyl)phenyl]pyridine
Formula:
C₁₂H₁₁NO MW: 185,222 g/mol Melting Pt: 172…176 °C Storage Temperature: Ambient |
MDL Number:
MFCD04114576 CAS Number: 217192-22-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...