4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine, N1-BOC protected 95%

Supplier: Apollo Scientific
Warning

OR14349-1G OR14349-10G OR14349-5G
APOSOR14349-1GEA 35 EUR
APOSOR14349-1G APOSOR14349-10G APOSOR14349-5G
4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine, N1-BOC protected 95%
4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazine, N1-BOC protected
Formula: C₂₁H₃₃BN₂O₄
MW: 388,309 g/mol
Melting Pt: 157…159 °C
Storage Temperature: Ambient
MDL Number: MFCD06411544
CAS Number: 470478-90-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR