{3-[(6-Methylpyrazin-2-yl)oxy]phenyl}methylamine 90%
Supplier: Apollo Scientific
|
Danger
|
OR14858-1G
OR14858-250MG
APOSOR14858-1GEA
379
EUR
APOSOR14858-1G
APOSOR14858-250MG
{3-[(6-Methylpyrazin-2-yl)oxy]phenyl}methylamine 90%
{3-[(6-Methylpyrazin-2-yl)oxy]phenyl}methylamine
Formula:
C₁₂H₁₃N₃O MW: 215,251 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09817526 CAS Number: 941716-89-8 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...