3-(3-Methyl-1,2,4-oxadiazol-5-yl)benzaldehyde 97%

Supplier: Apollo Scientific
Warning

Synonyms: 5-(3-Formylphenyl)-3-methyl-1,2,4-oxadiazole

OR14886-1G OR14886-250MG OR14886-100MG
APOSOR14886-1GEA 352 EUR
APOSOR14886-1G APOSOR14886-250MG APOSOR14886-100MG
3-(3-Methyl-1,2,4-oxadiazol-5-yl)benzaldehyde 97%
3-(3-Methyl-1,2,4-oxadiazol-5-yl)benzaldehyde
Formula: C₁₀H₈N₂O₂
MW: 188,183 g/mol
Melting Pt: 105…106 °C
Storage Temperature: Ambient
MDL Number: MFCD09817468
CAS Number: 273727-50-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR