5-(Pyrrolidin-1-yl)pyridine-2-carboxylic acid 95%

Supplier: Apollo Scientific
Warning

Synonyms: 1-(6-Carboxypyridin-3-yl)pyrrolidine, 2-Carboxy-5-(pyrrolidin-1-yl)pyridine

OR15955-1G OR15955-250MG
APOSOR15955-1GEA 433 EUR
APOSOR15955-1G APOSOR15955-250MG
5-(Pyrrolidin-1-yl)pyridine-2-carboxylic acid 95%
5-(Pyrrolidin-1-yl)pyridine-2-carboxylic acid
Formula: C₁₀H₁₂N₂O₂
MW: 192.214 g/mol
Melting Pt: 193.5…197 °C
Storage Temperature: Ambient
MDL Number: MFCD09879903
CAS Number: 950603-19-7
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR