Benzo[b]furan-3-boronic acid, pinacol ester 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-benzofuran, 2-(Benzofuran-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(1-Benzofuran-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR16510-250MG OR16510-1G OR16510-5G
APOSOR16510-250MGEA 258 EUR
APOSOR16510-250MG APOSOR16510-1G APOSOR16510-5G
Benzo[b]furan-3-boronic acid, pinacol ester 98%
Benzo[b]furan-3-boronic acid, pinacol ester
Formula: C₁₄H₁₇BO₃
MW: 244,094 g/mol
Melting Pt: 68…70 °C
Storage Temperature: Ambient
MDL Number: MFCD11841067
CAS Number: 796851-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR