4-(Benzyloxy)-5-methoxy-2-nitrobenzoic acid

Supplier: Apollo Scientific
Warning

OR17006-1G OR17006-250MG
APOSOR17006-1GEA 53.9 EUR
APOSOR17006-1G APOSOR17006-250MG
4-(Benzyloxy)-5-methoxy-2-nitrobenzoic acid
4-(Benzyloxy)-5-methoxy-2-nitrobenzoic acid
Formula: C₁₅H₁₃NO₆
Melting Pt: 192 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD04114356
CAS Number: 60547-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR