1-Methyl-6-phenyl-1H-imidazo[4,5-b]pyridin-2-amine 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-Amino-1-methyl-6-phenyl-4-azabenzimidazole, 2-Amino-1-methyl-6-phenylimidazo[4,5-b]pyridine
OR1700T-10MG
OR1700T-25MG
APOSOR1700T-10MGEA
147
EUR
APOSOR1700T-10MG
APOSOR1700T-25MG
1-Methyl-6-phenyl-1H-imidazo[4,5-b]pyridin-2-amine 98%
1-Methyl-6-phenyl-1H-imidazo[4,5-b]pyridin-2-amine
Formula:
C₁₃H₁₂N₄ MW: 224,261 g/mol Melting Pt: 227…229 °C Storage Temperature: Ambient |
MDL Number:
MFCD00210745 CAS Number: 105650-23-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...