1,2,3,6-Tetrahydropyridine-4-boronic acid, N-BOC protected 97%
Supplier: Apollo Scientific
Synonyms:
1-(tert-Butoxycarbonyl)-1,2,3,6-tetrahydropyridine-4-boronic acid, (1-(tert-Butoxycarbonyl)-1,2,3,6-tetrahydropyridin-4-yl)boronic acid
OR17710-1G
OR17710-5G
APOSOR17710-1GEA
134
EUR
APOSOR17710-1G
APOSOR17710-5G
1,2,3,6-Tetrahydropyridine-4-boronic acid, N-BOC protected 97%
1,2,3,6-Tetrahydropyridine-4-boronic acid, N-BOC protected
Formula:
C₁₀H₁₈BNO₄ MW: 227.065 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09997795 CAS Number: 844501-00-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...