4-Amino-5,7-dichloro-2,1,3-benzothiadiazole

Supplier: Apollo Scientific
Warning

Synonyms: 5,7-Dichloro-2,1,3-benzothiadiazol-4-amine

OR17856-100MG OR17856-1G OR17856-250MG
APOSOR17856-100MGEA 176 EUR
APOSOR17856-100MG APOSOR17856-1G APOSOR17856-250MG
4-Amino-5,7-dichloro-2,1,3-benzothiadiazole
4-Amino-5,7-dichloro-2,1,3-benzothiadiazole
Formula: C₆H₃Cl₂N₃S
MW: 220,079 g/mol
Melting Pt: 168…173 °C
Storage Temperature: Ambient
MDL Number: MFCD01118747
CAS Number: 16407-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR