3-Chloro-1H-pyrazole 95%

Supplier: Apollo Scientific
Warning

OR183405-1G OR183405-5G
APOSOR183405-1GEA 31.9 EUR
APOSOR183405-1G APOSOR183405-5G
3-Chloro-1H-pyrazole 95%
3-Chloro-1H-pyrazole
Formula: C₃H₃ClN₂
MW: 102,522 g/mol
Boiling Pt: 76…77 °C (0,5 mm Hg)
Melting Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD12756597
CAS Number: 14339-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR