5-Bromo-6-chloropicolinic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Bromo-6-chloropyridine-2-carboxylic acid
OR183415-250MG
OR183415-1G
OR183415-100MG
APOSOR183415-250MGEA
109
EUR
APOSOR183415-250MG
APOSOR183415-1G
APOSOR183415-100MG
5-Bromo-6-chloropicolinic acid
5-Bromo-6-chloropicolinic acid
Formula:
C₆H₃BrClNO₂ MW: 236.451 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09910220 CAS Number: 959958-25-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...