6-Bromo-5-chloropicolinic acid 95%
Supplier: Apollo Scientific
|
Warning
|
OR183417-5G
OR183417-1G
APOSOR183417-5GEA
521
EUR
APOSOR183417-5G
APOSOR183417-1G
6-Bromo-5-chloropicolinic acid 95%
6-Bromo-5-chloropicolinic acid
Formula:
C₆H₃BrClNO₂ MW: 236,451 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13185790 CAS Number: 1214328-42-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...