Methyl 3-amino-6-chlorobenzothiophene-2-carboxylate

Supplier: Apollo Scientific

Synonyms: Methyl 3-amino-6-chlorobenzo[b]thiophene-2-carboxylate

OR183425-1G OR183425-250MG
APOSOR183425-1GEA 659 EUR
APOSOR183425-1G APOSOR183425-250MG
Methyl 3-amino-6-chlorobenzothiophene-2-carboxylate
Methyl 3-amino-6-chlorobenzothiophene-2-carboxylate
Formula: C₁₀H₈ClNO₂S
MW: 241,694 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12032271
CAS Number: 35212-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR