Methyl 3-amino-5-iodobenzothiophene-2-carboxylate
Supplier: Apollo Scientific
OR183428-1G
OR183428-250MG
APOSOR183428-1GEA
364
EUR
APOSOR183428-1G
APOSOR183428-250MG
Methyl 3-amino-5-iodobenzothiophene-2-carboxylate
Methyl 3-amino-5-iodobenzothiophene-2-carboxylate
Formula:
C₁₀H₈INO₂S MW: 333,145 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29163655 CAS Number: 2090987-68-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...