4-(Pyrrolidin-1-yl)benzene-1,2-diamine
Supplier: Apollo Scientific
OR183526-1G
OR183526-250MG
APOSOR183526-1GEA
551
EUR
APOSOR183526-1G
APOSOR183526-250MG
4-(Pyrrolidin-1-yl)benzene-1,2-diamine
4-(Pyrrolidin-1-yl)benzene-1,2-diamine
Formula:
C₁₀H₁₅N₃ MW: 177,246 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11848945 CAS Number: 289913-84-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...