4-(4-Benzylpiperazin-1-yl)benzene-1,2-diamine

Supplier: Apollo Scientific

OR183527-1G OR183527-250MG
APOSOR183527-1GEA 607 EUR
APOSOR183527-1G APOSOR183527-250MG
4-(4-Benzylpiperazin-1-yl)benzene-1,2-diamine
4-(4-Benzylpiperazin-1-yl)benzene-1,2-diamine
Formula: C₁₇H₂₂N₄
MW: 282,383 g/mol
Storage Temperature: Ambient
CAS Number: 124802-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR