2-Bromo-3,5-dimethylpyridine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Bromo-3,5-lutidine
OR18610-1G
OR18610-5G
APOSOR18610-1GEA
32.1
EUR
APOSOR18610-1G
APOSOR18610-5G
2-Bromo-3,5-dimethylpyridine
2-Bromo-3,5-dimethylpyridine
Formula:
C₇H₈BrN MW: 186,049 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00234308 CAS Number: 92992-85-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...