2-{[(2-Ethylhexyl)oxy]carbonyl}benzoic acid 95%

Supplier: Apollo Scientific
Warning

Synonyms: Phthalic acid mono-2-ethylhexyl ester, Mono(2-ethylhexyl) phthalate, 2-(((2-Ethylhexyl)oxy)carbonyl)benzoic acid

OR19495-1G OR19495-5G
APOSOR19495-1GEA 129 EUR
APOSOR19495-1G APOSOR19495-5G
2-{[(2-Ethylhexyl)oxy]carbonyl}benzoic acid 95%
2-{[(2-Ethylhexyl)oxy]carbonyl}benzoic acid
Formula: C₁₆H₂₂O₄
MW: 278,343 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00041500
CAS Number: 4376-20-9
EINECS: 224-477-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR