4-Bromo-3-(bromomethyl)benzoic acid

Supplier: Apollo Scientific
Danger

Synonyms: 2-Bromo-5-carboxybenzyl bromide, α,4-Dibromo-m-toluic acid

OR19516-1G OR19516-250MG OR19516-5G
APOSOR19516-1GEA 279 EUR
APOSOR19516-1G APOSOR19516-250MG APOSOR19516-5G
4-Bromo-3-(bromomethyl)benzoic acid
4-Bromo-3-(bromomethyl)benzoic acid
Formula: C₈H₆Br₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD18378548
CAS Number: 887757-31-5
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR