3-Iodostyrene
Supplier: Apollo Scientific
|
Warning
|
OR19553-1G
OR19553-250MG
APOSOR19553-1GEA
348
EUR
APOSOR19553-1G
APOSOR19553-250MG
3-Iodostyrene
3-Iodostyrene
Formula:
C₈H₇I Boiling Pt: 71…73 °C (3 mm Hg) Density: 1,674 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD28103326 CAS Number: 4840-92-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...