2,4,5-Trichlorobenzoyl chloride
Supplier: Apollo Scientific
|
Danger
|
OR200112-1G
OR200112-250MG
APOSOR200112-1GEA
284
EUR
APOSOR200112-1G
APOSOR200112-250MG
2,4,5-Trichlorobenzoyl chloride
2,4,5-Trichlorobenzoyl chloride
Formula:
C₇H₂Cl₄O MW: 243,902 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18400116 CAS Number: 42221-49-8 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...