2-Amino-6-thien-2-ylmethyl)pyrimidin-4-ol

Supplier: Apollo Scientific

OR200137-1G OR200137-50MG OR200137-250MG OR200137-500MG
APOSOR200137-1GEA 510 EUR
APOSOR200137-1G APOSOR200137-50MG APOSOR200137-250MG APOSOR200137-500MG
2-Amino-6-thien-2-ylmethyl)pyrimidin-4-ol
2-Amino-6-thien-2-ylmethyl)pyrimidin-4-ol
Formula: C₉H₉N₃OS
MW: 207,252 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28154659
CAS Number: 1823494-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR