3-(Pyridin-3-yl)propanoyl chloride

Supplier: Apollo Scientific

OR200165-1G OR200165-250MG OR200165-500MG OR200165-2.5G
APOSOR200165-1GEA 589 EUR
APOSOR200165-1G APOSOR200165-250MG APOSOR200165-500MG APOSOR200165-2.5G
3-(Pyridin-3-yl)propanoyl chloride
3-(Pyridin-3-yl)propanoyl chloride
Formula: C₈H₈ClNO
MW: 169.608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02094016
CAS Number: 152656-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR