2-(4-Methylphenoxy)pyridine-3-carbonyl chloride

Supplier: Apollo Scientific
Danger

OR21707-1G OR21707-250MG
APOSOR21707-1GEA 557 EUR
APOSOR21707-1G APOSOR21707-250MG
2-(4-Methylphenoxy)pyridine-3-carbonyl chloride
2-(4-Methylphenoxy)pyridine-3-carbonyl chloride
Formula: C₁₃H₁₀ClNO₂
MW: 247,677 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00067838
CAS Number: 175135-79-2
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR