Diphenyl(vinyl)phosphine oxide
Supplier: Apollo Scientific
|
Warning
|
OR22135-1G
OR22135-5G
OR22135-10G
APOSOR22135-1GEA
109
EUR
APOSOR22135-1G
APOSOR22135-5G
APOSOR22135-10G
Diphenyl(vinyl)phosphine oxide
Diphenyl(vinyl)phosphine oxide
Formula:
C₁₄H₁₃OP Melting Pt: 115…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00114913 CAS Number: 2096-78-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...