6-Hydroxypyridazine-3-carboxaldehyde 90%, tech

Supplier: Apollo Scientific
Warning

Synonyms: 6-Formylpyridazin-3-ol, 3-Formyl-6-hydroxypyridazine, 6-Oxo-1,6-dihydropyridazine-3-carbaldehyde

OR2217-250MG
APOSOR2217-250MGEA 136 EUR
APOSOR2217-250MG
6-Hydroxypyridazine-3-carboxaldehyde 90%, tech
6-Hydroxypyridazine-3-carboxaldehyde
Formula: C₅H₄N₂O₂
MW: 124,097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09998728
CAS Number: 933734-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR