9-Oxodec-2-enoic acid 95%

Supplier: Apollo Scientific
Danger

OR22209-1G OR22209-250MG
APOSOR22209-1GEA 432 EUR
APOSOR22209-1G APOSOR22209-250MG
9-Oxodec-2-enoic acid 95%
9-Oxodec-2-enoic acid
Formula: C₁₀H₁₆O₃
Storage Temperature: Freezer
MDL Number: MFCD00204505
CAS Number: 334-20-3
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR