5-Phenyl-1,3-oxazole-4-carbonyl chloride

Supplier: Apollo Scientific
Danger

Synonyms: 4-(Chlorocarbonyl)-5-phenyl-1,3-oxazole

OR23138-1G OR23138-250MG
APOSOR23138-1GEA 778 EUR
APOSOR23138-1G APOSOR23138-250MG
5-Phenyl-1,3-oxazole-4-carbonyl chloride
5-Phenyl-1,3-oxazole-4-carbonyl chloride
Formula: C₁₀H₆ClNO₂
Melting Pt: 68…70 °C
Storage Temperature: Ambient
MDL Number: MFCD02681994
CAS Number: 337508-64-2
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR