[4-(Thien-2-yl)phenyl]methylamine

Supplier: Apollo Scientific
Danger

Synonyms: 2-[4-(Aminomethyl)phenyl]thiophene, 4-(Thien-2-yl)benzylamine

OR23166-1G OR23166-25G OR23166-5G
APOSOR23166-1GEA 94.4 EUR
APOSOR23166-1G APOSOR23166-25G APOSOR23166-5G
[4-(Thien-2-yl)phenyl]methylamine
[4-(Thien-2-yl)phenyl]methylamine
Formula: C₁₁H₁₁NS
MW: 189.277 g/mol
Melting Pt: 76…77 °C
Storage Temperature: Ambient
MDL Number: MFCD02682030
CAS Number: 203436-48-0
UN: 3259
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR