6-(Morpholin-4-yl)pyridine-3-sulphonyl chloride 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
5-(Chlorosulphonyl)-2-(morpholin-4-yl)pyridine, 4-[5-(Chlorosulphonyl)pyridin-2-yl]morpholine
OR23272-1G
OR23272-250MG
APOSOR23272-1GEA
352
EUR
APOSOR23272-1G
APOSOR23272-250MG
6-(Morpholin-4-yl)pyridine-3-sulphonyl chloride 97%
6-(Morpholin-4-yl)pyridine-3-sulphonyl chloride
Formula:
C₉H₁₁ClN₂O₃S MW: 262,713 g/mol Melting Pt: 118 °C Storage Temperature: Ambient |
MDL Number:
MFCD02681929 CAS Number: 337508-68-6 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...